missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT and CytPix Waste BTL Tubing

Catalog No. 502723657
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502723657 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502723657 Supplier Invitrogen™ Supplier No. 100096710CST

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This is replacement tubing for the waste bottle of the Attune NxT Acoustic Focusing Cytometer

  • This is replacement tubing for the waste bottle of the Attune NxT Acoustic Focusing Cytometer.
  • The tubing can be replaced easily without replacing the entire bottle assembly if the tubing connector to the instrument is broken.

Specifications

Product Line FPL
Quantity Each
Format Tubing
Shipping Condition Room Temperature
For Use With (Equipment) Attune NxT
Product Type Tubing