Learn More
Invitrogen™ Attune™ NxT Violet 6 Conversion Kit
The Attune NxT Violet 6 Conversion kits are designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet.
Supplier:  Invitrogen™ A36571
 View more versions of this product
Catalog No. A36571
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Attune NxT Violet 6 Conversion kits are for use in the reconfiguration of four-channel Attune NxT instruments to enable detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out in your laboratory by a Field Service Engineer.
Specifications
Chemical Identifiers
Attune
Room Temperature
Conversion Kit
Optical block and filters
Attune NxT Four-Channel Violet Instrument
Specifications
Attune
1 kit
Room Temperature
Conversion Kit
Optical block and filters
Red
Attune NxT Four-Channel Violet Instrument
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.