missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Software, single license

Catalog No. A25554
Change view
Click to view available options
Quantity:
1 license
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25554 1 license
501034982 1 license
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25554 Supplier Invitrogen™ Supplier No. LSA25554

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Attune NxT Software, single license

Attune™ NxT Software is used to operate and maintain the Attune™ NxT acoustic focusing cytometer. It enables powerful user-defined experimental analysis via an intuitive, easy-to-use interface and provides advanced tools for both acquisition and analysis that are simple enough for users at all experience levels.

  • Simplified use—streamlined, intuitive interface
  • Speed—faster real-time analysis of large data sets
  • Easy instrument operation—automated maintenance and instrument performance monitoring

    Simplified use
    Attune™ NxT software employs a menu system that presents the features you need when you need them without a lot of scrolling and clicking. The customization tab allows you to pre-define settings for plots, gates, experiments, reports, and overlays. To simplify experimental set up, the preview screen shows all plot combinations instantaneously allowing you to easily add them to the experiment workspace. Compensation is automated or user-defined and can be set up using a compensation guide.

    Speed increases productivity
    Attune™ NxT Software has been designed to maximize the efficiency in performing data analysis. It uses a Windows™ 7 64-bit system to provide fast refresh rates for large data sets of up to 20 million events created by multi-laser, multi-parameter instrumentation. The software allows you to immediately visualize changes to your data plots in real-time as you make adjustments. This allows you to spend less time analyzing data and more time on your research.

    Easy instrument operation
    Flow cytometers often require the user to spend time ensuring the quality of the system through maintenance tasks and proper QC, since changes in lasers, fluidics, and optics can affect the quality of the data generated. Attune™ NxT Software takes many of these manual tasks and fully automates them to help reduce time spent on instrument maintenance. The software provides an automated quality control instrument tracking system, important for collecting and analyzing accurate data requiring minimal user interaction.

    Software features:
    Instrument controls
    Automated maintenance and performance testing
    Data collection
    Visualization tools
    Dot plot, density plot, histogram
    Histogram marker, quad marker
    Customized logic gates
    Data plot preview for quick plot set-up
    Data display—log, linear, log transforms
    Statistics—mean, median, mode, %CV, rCV, SD, % total, % gated
    Experiment set-up
    Automated compensation
    Auto layout and freeform workspace layouts
    Analysis tools
    Multi-file selection tools
    File management
    Import/export of files and experiments
    Files stored as FCS3.1 format
    User management & security
    User and administrator accounts—basic/admin
  • Specifications

    Product Line Attune
    Content And Storage 1 Attune™ NxT Software dongle, 1 copy Attune™ NxT Software
    Quantity 1 license
    Shipping Condition Room Temperature
    Software Type 1 individual user license
    For Use With (Equipment) Attune NxT Acoustic Focusing Cytometer
    Product Type Software

    For Research Use Only. Not for use in diagnostic procedures.