missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT and CytPix Shutdown Bottle

Catalog No. 501034994
Encompass
Change view
Click to view available options
Quantity:
1 Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501034994 1 Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501034994 Supplier Invitrogen™ Supplier No. 100022154

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This is a 175 mL replacement bottle for shutdown solution for the Attune™ NxT Acoustic Focusing Cytometer.

This is a 175 mL replacement bottle for shutdown solution for the Attune™ NxT Acoustic Focusing Cytometer.

Specifications

Product Line Attune
Content And Storage Store at room temperature.
Quantity 1 Bottle
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Shutdown Bottle

For Research Use Only. Not for use in diagnostic procedures.