missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT and CytPix Sample Syringe

Catalog No. 501035012
Encompass
Change view
Click to view available options
Quantity:
1 Syringe
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501035012 1 Syringe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501035012 Supplier Invitrogen™ Supplier No. 100022591

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This 1 mL sample syringe is used with the fluidics system of the Attune™ NxT Acoustic Focusing Cytometer to move sample to the flow cell.

This 1 mL sample syringe is used with the fluidics system of the Attune™ NxT Acoustic Focusing Cytometer to move sample to the flow cell. Replacement of this syringe is recommended as part of instrument maintenance.

Specifications

Product Line Attune
Content And Storage Store at room temperature.
Quantity 1 Syringe
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Sampler Syringe

For Research Use Only. Not for use in diagnostic procedures.