missing translation for 'onlineSavingsMsg'
Learn More

Attune™ NxT and CytPix Maintenance Kit

Catalog No. A43038
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A43038 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A43038 Supplier Thermo Fisher Scientific Supplier No. A43038

The Attune NxT Maintenance Kit will help you avoid down-time and ensure optimal operation of your Attune NxT Acoustic Focusing Cytometer hardware.

This convenient kit includes sufficient supplies to perform all recommended preventative maintenance procedures for 12 months. These procedures are: replacement of the syringe (bi-yearly), SIP tube (yearly), and instrument bottles (yearly), as well as performance of system decontamination (quarterly). The kit includes 1 waste bottle, 1 focusing fluid tank, 12 focusing fluid filters, 2 syringes, 1 SIP tube, and 1 debubble solution.

Specifications

Product Line Attune
Content And Storage • 1 waste bottle
• 1 focusing fluid bottle
• 12 focusing fluid filters
• 2 syringes
• 1 SIP
• 1 debubble solution

Store at room temperature.

Quantity 1 kit
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Block Format Non-Interchangeable
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Maintenance Kit
How often should I change filters for the Attune NxT Acoustic Focusing Cytometer?

Filters are recommended for replacement once every three months, unless performing small particle detection when once-a-month replacement is recommended.

Can I purchase the individual components of the Attune NxT Maintenance Kit as standalone products?

Yes. Here are the catalog numbers for the individual components:
- Cat. No. 100022156: 1 Waste bottle
- Cat. No. 100022155: 1 Focusing fluid bottle
- Cat. No. 100022587: 12 Focusing fluid filters
- Cat. No. 100022591: 2 Syringes
- Cat. No. 4490099: 1 SIP
- Cat. No. A10496: Debubble solution

Is the debubble solution provided in the Attune NxT Maintenance Kit the same as the standalone product, Attune Debubble Solution (1X), Cat. No. A10496?

Yes. It is the same 50 mL bottle of 1X Debubble Solution as provided in Attune Debubble Solution (1X).

For Research Use Only. Not for use in diagnostic procedures.