missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT and CytPix Focus BTL Tubing

Catalog No. 502723656
Change view
Click to view available options
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502723656 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502723656 Supplier Invitrogen™ Supplier No. 100096711CST

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This is replacement tubing for the focusing fluid bottle of the Attune NxT Acoustic Focusing Cytometer

  • This is replacement tubing for the focusing fluid bottle of the Attune NxT Acoustic Focusing Cytometer.
  • The tubing can be replaced easily without replacing the entire bottle assembly if the tubing connector to the instrument is broken.

Specifications

Product Line FPL
Includes 1 tubing set
Quantity 1 each
Format Tubing
Shipping Condition Room temperature
For Use With (Equipment) Attune NxT
Product Type Tubing