missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT and CytPix Focus Bottle

Catalog No. 502723660
Change view
Click to view available options
Quantity:
1 bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
502723660 1 bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 502723660 Supplier Invitrogen™ Supplier No. 100085929CST

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This is a 1.9-L replacement bottle for the focusing fluid tank on the Attune NxT Acoustic Focusing Cytometer.

  • This is a 1.9-L replacement bottle for the focusing fluid tank on the Attune NxT Acoustic Focusing Cytometer.
  • The bottle is designed for easy detachment from the instrument for refill and with more robust fittings to prevent breakage.
  • This bottle replaces discontinued Cat. No. 100022155.

Specifications

Product Type Flow Cytometer Accessory
Quantity 1 bottle
Content And Storage 1 Bottle
Product Line FPL
Shipping Condition Room temperature
Format Bottle
For Use With (Equipment) Attune NxT