Learn More
Invitrogen™ Attune™ NxT External Fluid Supply
Attune NxT External Fluid Supply
Supplier: Invitrogen™ A28006
Description
The Attune™ NxT External Fluid Supply is an accessory for the Attune™ NxT Acoustic Focusing Cytometer. It was designed to facilitate the use of a larger buffer and waste container. The system works by monitoring and supplying the internal fluid tanks on the Attune NxT instrument so that the system can run without having to stop instrument operation to change out the fluid tanks or empty the waste. The external fluid system uses the 10 L cube of Attune™ Focusing Fluid (Cat. No. A24904) and a Nalgene™ waste bottle. The external fluid supply is ideal for use with the Attune™ NxT Auto Sampler and enables the collection of approximately 40 96-well or 10 384-well plates without changing the fluid tanks.Spécifications
| Flow Cytometer Accessory | |
| Room Temperature | |
| Attune™ NxT Acoustic Focusing Cytometer | |
| Attune |
For Research Use Only. Not for use in diagnostic procedures.