missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Automation Bundle, stacks/hotel

Catalog No. A35220
Change view
Click to view available options
Quantity:
1 Bundle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A35220 1 Bundle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A35220 Supplier Invitrogen™ Supplier No. A35220

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Automation robot that delivers improved walk-away time and smarter productivity

  • Automate your flow cytometry workstation with the Attune NxT and Orbitor RS2 Automation Bundle. The Orbitor RS2 Microplate Mover is an automation robot that delivers improved walk-away time and smarter productivity. This Automation Bundle is designed for an Attune NxT Acoustic Focusing Cytometer integrated with the Attune NxT, CytKick, or CytKick Max autosampler.
  • To address your plate format storage and handling, there are three different configurations: one with three sequential access stacks (40 plates per stack) (Cat. No. A33007), one with three random access hotels (24 plates/3 hotels) (Cat. No. A33008), and one with two sequential access stacks and one random access hotel (this page).
  • Extensive vertical reach allows the Orbitor RS2 Microplate Mover to access virtually any microplate instrument location, including stacked instruments, and its telescoping arm can reach a variety of distances. The ingenious arm travels through the base, resulting for rapid and efficient movements. This robot has a full 360°workspace with unlimited base rotation; this eliminates the need for hard stops and reverse movements.
  • Advanced closed-loop motion control provides collision detection and position monitoring, allowing the Orbitor RS2 Microplate Mover to quickly resume without the need to re-home. With a wide range of grip-control functionality, this robot combines performance with innovation. From variable force control to plate detection, the feature set is designed to impress. The result is remarkable speed, precise motion, and consistent, accurate plate placement.
  • The Orbitor RS2 Microplate Mover is powered by Thermo Scientific Momentum Integration Software with features such as Momentum scheduling software, error detection and recovery, and a dynamic scheduler that allows changes while running.

Specifications

Product Line Attune
Content And Storage • 1 Orbitor RS2 Microplate Mover
• Integrated barcode reader
• 1 Momentuum Software License
• 2 plate stacks
• 1 plate hotel

Quantity 1 Bundle
Shipping Condition Room Temperature
Block Format Non-Interchangeable
For Use With (Equipment) Attune NxT Acoustic Focusing Cytometer
Product Type Automation Bundle

For Research Use Only. Not for use in diagnostic procedures.