Learn More
Invitrogen™ Attune™ Laser Upgrade Kits
Description
Enhance the capabilities of your Attune NxT Acoustic Focusing Flow Cytometer and Attune CytPix Imaging Enhanced Flow Cytometer with our versatile laser upgrade kits. Available in red, yellow, violet, and green options, these upgrade kits are designed to expand your instrument's fluorescence detection capabilities, enabling a broader range of research applications and enhancing the versatility of your flow cytometry experiments.
Expanded capabilities
Maximize your research potential with our laser upgrade kits. Each kit is tailored to specific excitation wavelengths, allowing you to utilize a variety of labels, dyes, and applications. For example, the Red Laser Upgrade Kit expands the 637-nm excitation capabilities, facilitating the detection of APC, Alexa Fluor 700, APC-tandem dyes, and functional reagents such as CellTrace Far Red dye, LIVE/DEAD Fixable far red or near IR dyes, Vybrant DyeCycle Ruby dye, and far red–exciting fluorescent proteins. Similar enhancements are available with the yellow, violet, and green laser upgrade kits, each broadening the scope of detectable fluorophores and applications.
Cost-effective solutions
Upgrading your existing flow cytometer is a cost-effective way to extend its capabilities without the need to invest in new instruments. These upgrades allow you to configure your Attune NxT and Attune CytPix flow cytometers with different laser configurations, meeting the evolving demands of your research while optimizing your budget.
Simple installation
The Attune NxT and Attune CytPix flow cytometers are designed for easy upgrades. Each laser upgrade kit includes all necessary optical components and can be installed by a qualified service engineer in typically less than four hours. This minimizes downtime and ensures that your instrument is quickly back in operation with enhanced functionality.
Specifications
Specifications
| Product Line | Attune |
| Content And Storage | Contains violet laser with fiber optic cable and optics block. |
| Quantity | 1 Kit |
| Color | Violet |
| For Use With (Equipment) | Attune Acoustic Focusing Cytometer |
| Product Type | Laser Upgrade Kit |
For Research Use Only. Not for use in diagnostic procedures.