missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Attune™ Debubble Solution (1X)
Description
Attune™ Debubble Solution is used to remove excess air bubbles that may develop in the fluidic lines of the Attune™ NxT Acoustic Focusing Cytometer.
Minimizes bubble formation
Attune™ Debubble Solution has been formulated to minimize bubble formation that may occur in the system lines overnight or after system maintenance. This solution is used during the automated Debubble function of the Attune™ acoustic focusing cytometer. It is also used after the regular quarterly maintenance/system decontamination function to help ensure that all air introduced during this cleaning process is removed prior to analyzing samples on the system.
Convenient
This ready-to-use 1X debubble solution was designed to eliminate the need for dilution prior to use and the need for access to a purified water system.
High quality
Each batch is traceable by its lot number designation and has been validated for use on the Attune™ acoustic focusing cytometer.
Minimizes bubble formation
Attune™ Debubble Solution has been formulated to minimize bubble formation that may occur in the system lines overnight or after system maintenance. This solution is used during the automated Debubble function of the Attune™ acoustic focusing cytometer. It is also used after the regular quarterly maintenance/system decontamination function to help ensure that all air introduced during this cleaning process is removed prior to analyzing samples on the system.
Convenient
This ready-to-use 1X debubble solution was designed to eliminate the need for dilution prior to use and the need for access to a purified water system.
High quality
Each batch is traceable by its lot number designation and has been validated for use on the Attune™ acoustic focusing cytometer.
Specifications
Specifications
| Product Line | Attune |
| Quantity | 50 mL |
| For Use With (Equipment) | Attune™ Acoustic Focusing Cytometer |
| Content And Storage | Store at room temperature. |
| Product Type | Debubble Solution |
For Research Use Only. Not for use in diagnostic procedures.