missing translation for 'onlineSavingsMsg'
Learn More

Atto 590 NHS ester suitable for Fluorescence, BioReagent, 60 (coupling to amines), ≥60% (coupling to amines), MilliporeSigma™ Supelco™

Catalog No. 111015124
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015124 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015124 Supplier Merck Emd Millipore Supplier No. 796361MGF

Chemical Identifiers

MDL Number MFCD03456133

Specifications

CAS 670269-33-7
Linear Formula C41H42 ClN3O11
Quantity 1 mg
Formula Weight 788.24
Percent Purity ≥60% (coupling to amines)

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.