missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium Trioxide, Crystal, ACS, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystals or Needles |
| Quantity | 45 kg |
| Molecular Weight (g/mol) | 99.99 |
| Percent Purity | 98% |
| Grade | ACS/Reagent |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | Chromium Trioxide, Crystal |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.