missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atrolactic Acid Hydrate 98.0+%, TCI America™
$223.68 - $846.29
Chemical Identifiers
| CAS | 515-30-0 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00004451 |
| InChI Key | NWCHELUCVWSRRS-UHFFFAOYSA-N |
| Synonym | atrolactic acid, 2-phenyllactic acid, 2-hydroxy-2-phenylpropionic acid, alpha-methylmandelic acid, phenyllactic acid, 2-phenyl-2-hydroxypropionic acid, dl-atrolactic acid, alpha-hydroxy-alpha-phenylpropionic acid, mandelic acid, .alpha.-methyl, benzeneacetic acid, .alpha.-hydroxy-.alpha.-methyl |
| PubChem CID | 1303 |
| ChEBI | CHEBI:50392 |
| IUPAC Name | 2-hydroxy-2-phenylpropanoic acid |
| SMILES | CC(C1=CC=CC=C1)(C(=O)O)O |
Chemical Identifiers
| 515-30-0 | |
| 166.176 | |
| NWCHELUCVWSRRS-UHFFFAOYSA-N | |
| 1303 | |
| 2-hydroxy-2-phenylpropanoic acid |
| C9H10O3 | |
| MFCD00004451 | |
| atrolactic acid, 2-phenyllactic acid, 2-hydroxy-2-phenylpropionic acid, alpha-methylmandelic acid, phenyllactic acid, 2-phenyl-2-hydroxypropionic acid, dl-atrolactic acid, alpha-hydroxy-alpha-phenylpropionic acid, mandelic acid, .alpha.-methyl, benzeneacetic acid, .alpha.-hydroxy-.alpha.-methyl | |
| CHEBI:50392 | |
| CC(C1=CC=CC=C1)(C(=O)O)O |
Specifications
| 515-30-0 | |
| White-Yellow | |
| MFCD00004451 | |
| atrolactic acid, 2-phenyllactic acid, 2-hydroxy-2-phenylpropionic acid, alpha-methylmandelic acid, phenyllactic acid, 2-phenyl-2-hydroxypropionic acid, dl-atrolactic acid, alpha-hydroxy-alpha-phenylpropionic acid, mandelic acid, .alpha.-methyl, benzeneacetic acid, .alpha.-hydroxy-.alpha.-methyl | |
| CC(C1=CC=CC=C1)(C(=O)O)O | |
| 166.176 | |
| CHEBI:50392 | |
| ≥98.0% (HPLC) | |
| Atrolactic Acid Hydrate |
| 91°C | |
| C9H10O3 | |
| 1 g | |
| NWCHELUCVWSRRS-UHFFFAOYSA-N | |
| 2-hydroxy-2-phenylpropanoic acid | |
| 1303 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes