missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP Solution (10 mM)
Description
ATP is an essential component for kinase assays using LanthaScreen™ Technology. Invitrogen's ATP is 10mM in water.
Specifications
Specifications
| Content And Storage | ATP is shipped on dry ice and should be stored at -20°C. |
| Assay | Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity |
| Shipping Condition | Dry Ice |
| Concentration | 10 mM |
| Label or Dye | Unlabeled |
| Quantity | 500 μL |
For Research Use Only. Not for use in diagnostic procedures.