missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ KV300 Aramid Fiber Cut-Resistant Gloves, Natural Rubber Coating

Catalog No. p-4505931
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19270234 Small
19270235 Medium
19270236 Large
19270237 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19270234 Supplier SHOWA™ Supplier No. KV300S07
Only null left

Gloves feature a 10-gauge, seamless knit liner with a latex coating for enhanced grip capabilities in wet or dry conditions.

  • Aramid fiber gloves provide ANSI Level A3 Cut Resistance
  • Rough-textured latex coating enhances wet and dry grip
  • Uncoated knuckles ensure flexibility and precision

Specifications

ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Latex
Color Yellow, Blue
Cut Resistance Rating ANSI A3
Exterior Finish Fully Textured
Hand Specific
Material Aramid Fiber
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:3X43CP
For Use With (Application) Heavy equipment, bottling, glass and windows, bottling and canning, police and defense purposes
Glove Cuff Style Knit
Model KV300-07
Puncture Rating ANSI/ISEA 105-16 Level 5
Series KV300
Yarn Gauge 10
Length (English) 9.5 in.
Length (Metric) 24.1 cm
Show More Show Less