missing translation for 'onlineSavingsMsg'
Learn More

Cesium Chloride, Optical Grade, 99%, Spectrum™ Chemical

Catalog No. 18609627
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609627 25 kg Amber Glass Bottle
18609622 25 g Amber Glass Bottle
18609623 100 g Amber Glass Bottle
18609624 500 g Amber Glass Bottle
18609625 1 kg Amber Glass Bottle
18609626 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18609627 Supplier Spectrum Chemical Mfg Cor Supplier No. CE11725KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7647-17-8 | CsCl | 168.37 g/mol

  • Suitable for isopycnic centrifugation and in research and development

Chemical Identifiers

CAS 7647-17-8

Specifications

Melting Point 645°C
Color White
CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 25 kg
Assay Percent Range 99%
Molecular Formula CsCl
Synonym Tricesium Trichloride, Cesium Monochloride, Dicesium Dichloride
Solubility Information Soluble in Water, Methanol, Ethanol
Formula Weight 168.37 g/mol
Percent Purity 99%
Grade Optical
Odor Odorless
Packaging Amber Glass Bottle
Appearance Colorless Crystal
Decomposition Information Cesium Oxides, Hydrogen Chloride
Density 3.988 g/cm³
Chemical Name or Material Cesium Chloride
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.