Learn More
Enzo Life Sciences ATF6 monoclonal antibody (70B1413.1) (100 g)
Supplier: Enzo ALX804381C100
Clone: 70B1413.1. Host: Mouse. Isotype: IgG1. Immunogen: Partialhuman ATF6 (activating transcription factor 6) containing aa 1-273. UniProt: P18850. Species Reactivity: Human, Mouse, Rat, Rabbit, Hamster. Applications: ICC, IHC (PS), IP, WB, ChIP. Recommended dilutions: Western Blot (1-5g/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.05% BSA and 0.05% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20C.