Learn More
Astrospec 3000™ Eyewear
Quality, comfort and styling make these a popular choice.
Supplier:  Customer Channels Group (Ccg) S1359EMD
 View more versions of this product
Catalog No. S476252
Only null left
Explore available promotions
Description
Description
  • Adjustable temple length and lens inclination system
  • Duoflex™ comfort cushions relieve pressure behind the ear
  • Optically correct lens has Uvextreme™ AF anti-fog and anti-scratch coating
Compliance
Meet ANSI™ Z87.1-2003 standards.
Specifications
Specifications
Scratch-resistant
Clear
Duoflex
Meet ANSI Z87.1-2003 standards.
Black
Product Suggestions
Product Suggestions