Learn More
Fisherbrand™ ASTM™ Hydrometers, Plain Form
API hydrometers are available in regular and ASTM™ recommended forms.
 View more versions of this product
Catalog No. 11603H
Only null left
Description
Description
These hydrometers are manufactured to meet or exceed ASTM™ E 100 specifications which are accepted by NIST™ and U.S. Bureau of Mines.
  • Permanently marked with individual serial numbers
  • Without thermometer in body
  • Approximate length: 330mm
  • Temperature of standardization: 60°F
Order Info
A certificate of calibration is available; simply add the letter C to the end of the catalog number when ordering.
Certifications
A certificate is provided to indicate instrument traceability to standards provided by NIST.
Compliance
Meet or exceed ASTM E 100 specifications which are accepted by NIST and U.S. Bureau of Mines, Calibrated according to ASTM E 126 specifications.
Specifications
Chemical Identifiers
0.1°
330 mm
60°F
Specifications
10H; Without 3-pt certification
NIST, Cited in ASTM E100-14 Table 1, ASTM E126
60°F
0.1°
330 mm
89 to 101° ASTM 10H-62
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Accessories