Learn More
Ritmed™ AssureWear™ SMMS VersaGown™ Isolation Gowns with Flexneck™ Technology
Catalog No. 19910748
Change view
Click to view available options
Size:
X-Large
Catalog No. Size
19910748 X-Large
1 options
Catalog No. 19910748 Supplier Ritmed™ Supplier No. A69912
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Disposable SMMS polypropylene isolation gowns are designed to protect health professionals and patients from cross infection. Patented Flexneck™ technology for fast and easy donning and removal.
  • Patented Flexneck™ technology allows fast and easy donning and removal while providing exceptional protection and comfort
  • Innovative Flexneck™ closure quickly and easily passes over the head to instantly provide complete coverage and eliminate the risk posed by untied gaping gowns that can slip and expose skin
  • Flexneck™ closure also allows easy doffing that allows gown to swiftly yet gently tear away from the body preventing self-contamination.
Specifications
Size X-Large
No. of Pockets None
Gender Unisex
Product Type Isolation Gown
Material SMMS Polypropylene
Color Yellow
Closure Type Over-the-Head
Disposable Single-use
Sterility Non-sterile
Cuff Style Elastic
Latex Free Yes
Model A69912
Sleeve Style Long Sleeve
Includes Flat Front Belt
Certifications/Compliance AAMI PB70 level 1, Medical Device Class 1
Autoclavable Not Autoclavable
Show More Show Less