Learn More
Medicom™ AssureWear™ VersaGown™ Chemotherapy Tested Gown, Flexneck™ Technology
Catalog No. 296367832
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296367832 X-Large
296367831 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 296367832 Supplier Medicom™ Supplier No. A69932
Polycoated gowns are chemotherapy tested and meet AAMI Level 3 standards.
  • VersaGown™ with Flexneck™ Plus technology delivers best-in-class infection control compliance.
  • Flexneck™ Plus technology has two double elastic closures: one that facilitates neck closure and a second that helps ensure that the back of the gown overlaps.
  • This tie-free design enables quick and easy donning and doffing while maintaining exceptional protection and comfort.
  • Poly-coated and with thumb loop cuffs to prevent sleeves from riding up and ensure complete wrist coverage under exam gloves.
  • Rated AAMI Level 3 (CAN ONLY)
  • 100/Cs.
Specifications
Product Type Chemotherapy Gown
Gender Unisex
No. of Pockets None
Size X-Large
Color Blue
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Sleeve Style Long Sleeve
Autoclavable Not Autoclavable
Show More Show Less