missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medicom™ AssureWear™ Sterile Surgical Gowns, AAMI Level 3
Professional surgical gown designed to prevent penetration and cross-contamination of body fluids between patients and healthcare providers.
Supplier: Medicom™ S67001
Description
- Provides breathable protection during sterile surgical procedures.
- Constructed from heavy weight, multi-layer SMS material for durability and fluid resistance.
- Features Velcro neck closure, knit cuffs, full back design, and secure side ties for optimal fit and coverage.
- AAMI Level 3-suitable for procedures with moderate fluid or blood exposure risk.
- Includes two 4-ply absorbent towels for added convenience.
- Latex-free to reduce the risk of allergic reactions.
Specifications
| Surgical Gown | |
| Unisex | |
| Spunbond Polypropylene | |
| Tape-tab at Neck | |
| Sterile | |
| Yes | |
| Not Autoclavable |
| None | |
| Large | |
| Blue | |
| Single-use | |
| Knit | |
| Long Sleeve |