Learn More
Medicom™ AssureWear™ Blue SMS Lab Coats
Catalog No. 19910812
Change view
Click to view available options
Size:
Medium
Large
X-Large
3X-Large
4X-Large
7X-Large
9X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19910812 9X-Large
19910805 Medium
19910806 Large
19910807 X-Large
19910809 3X-Large
19910810 4X-Large
19910811 7X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19910812 Supplier Medicom™ Supplier No. A8040
Non-woven SMS lab coat with knitted cuffs is designed to protect healthcare professionals, lab technicians, and other wearers from fluids and cross-contamination.
  • Non-woven 3-layer SMS
  • Knit cuffs
  • Knit collar
Specifications
Product Type General Purpose Lab Coat
Size 9X-Large
Gender Unisex
Material SMS
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Style Knee-length
No. of Pockets None
Collar Type Knit
Latex Free Yes
Model A8040
For Use With (Application) Healthcare, Laboratories
Show More Show Less