missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureWear Polyethylene Isolation Gown

Catalog No. 296366743
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
296366743 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296366743 Supplier Medicom™ Supplier No. MED8025

Isolation gown with an over-the-head design and thumb loop cuffs.

  • Designed to protect healthcare professionals and patients against cross-contamination
  • Comfortable fit
  • Thumb loop cuffs
  • Sewn seams provide superior strength
  • Single-use
  • Disposable and latex free
  • Spunbond polypropylene

Specifications

Product Type General Purpose Gown
Gender Unisex
No. of Pockets None
For Use With (Application) Protecting health professionals and patients from cross infection
Size X-Large
Material Polyethylene
Color Blue
Closure Type Over-the-Head
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Certifications/Compliance Non-AAMI
Autoclavable Not Autoclavable
Show More Show Less