Learn More
Medicom™ AssureWear™ Polyethylene Isolation Gown
Catalog No. 296366744
Change view
Click to view available options
Color:
Blue
Catalog No. Color
296366744 Blue
1 options
Catalog No. 296366744 Supplier Medicom™ Supplier No. 8026
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AssureWear™ Polyethylene Isolation Gown with over-the head design.
  • Spunbond polyethylene gown with over-the-head design and thumb loop cuffs.
  • Sewn seams provide superior strength.
  • Latex free.
  • 75/Cs.
  • Non AAMI rated
Specifications
Gender Unisex
No. of Pockets None
Size X-Large
Product Type Isolation Gown
Material Polyethylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Autoclavable Not Autoclavable
Show More Show Less