missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureWear™ White SMS Lab Coats

Catalog No. p-7238935
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19910793 Small
19910794 Medium
19910795 Large
19910796 X-Large
19910797 2X-Large
19910798 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19910793 Supplier Medicom™ Supplier No. A8041
Only null left

Non-woven SMS lab coat with knitted cuffs is designed to protect healthcare professionals, lab technicians, and other wearers from fluids and cross-contamination.

  • Non-woven 3-layer SMS
  • Knit cuffs
  • Knit collar

Specifications

Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material SMS
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Style Knee-length
No. of Pockets None
Collar Type Knit
Latex Free Yes
Model A8041
For Use With (Application) Healthcare, Laboratories
Autoclavable Not Autoclavable
Show More Show Less