missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ AssureTouch™ Agile™ Nitrile Medical Examination Gloves with Textured Fingertips

Catalog No. 17101085
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17101085 Medium
17101083 X-Small
17101084 Small
17101086 Large
17101087 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17101085 Supplier Medicom™ Supplier No. 9998C
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AssureTouch™ Agile™ Nitrile Medical Examination Gloves with Textured Fingertips. This nitrile glove offers optimal puncture and tear resistance without compromising dexterity or tactility. Textured fingertips ensure a secure grip, even when wet. Chemo tested.

  • These nitrile gloves offer superior puncture, tear and chemical resistance without compromising dexterity or tactility.
  • Textured fingertips ensure a secure grip, even when wet.
  • Chemo tested.

Specifications

Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free No
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 4.5 mil
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Medium
Sterility Non-sterile
Thickness (English) 4.5 mil
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model 9998-C
Quantity 1000/Cs.
Show More Show Less