Learn More
Medicom™ AssureMask Valiant™ Earloop Face Masks
Procedure earloop face masks provide ASTM Level 1, 2 or 3 protection as well as optimal comfort and breathability.
Supplier: Medicom™ 205717
Description
- Non-woven spunbond polypropylene outer layer provides maximum protection against fluid penetration
- High-quality media ensure optimal filtration and breathability
- Inner layer is made of dye-free non-woven spunbond polypropylene that will not lint or tear during use
- Ample-length earloops are soft, strong, free of natural rubber latex, ultrasonically sealed, and attached to the outside of the mask to prevent pulling and irritation
- Adjustable, enclosed, plastic-covered, iron nosepiece provides customized protection and comfort
- Shingle pleat design prevents fluid pooling and cross-contamination
- Made in North America to meet the highest quality and regulatory standards
- Available in assorted colors
Specifications
| Medical/Procedural Face Mask | |
| Spunbond Polypropylene | |
| Non-sterile | |
| Single-use | |
| Yes | |
| 50 Masks/Box, 10 Boxes/Cs. |
| Earloops | |
| Standard | |
| Pleated | |
| ≥95% (PFE), ≥95% (BFE) | |
| 205717 |