Learn More
Medicom™ AssureMask Precision™ Tie-On Face Masks
Tie-on face masks provide ASTM Level 1, 2 or 3 protection. Made from high quality materials that enhance breathability, these masks have soft, ample-length ties, an adjustable nose piece and a soft inner lining to ensure a comfortable, customized fit.
Supplier: Medicom™ 206814
Description
- Non-woven spunbond outer layer, provides maximum protection against fluid penetration
- Ample length ties are soft, strong, ultrasonically sealed and attached to the outside of the mask to prevent pulling and irritation
- Shingle pleat design prevents fluid pooling and cross-contamination
- High quality media ensures optimal filtration and breathability
- SureView™ Technology Fog-free nose strip ensures an unobstructed view of the operative field
Specifications
| Medical/Procedural Face Mask | |
| Spunbond Polypropylene | |
| Non-sterile | |
| Single-use | |
| Yes | |
| 50 Masks/Box, 6 Boxes/Cs. |
| Ties | |
| Standard | |
| Pleated | |
| ≥95% (PFE), ≥95% (BFE) | |
| 206814 |