Learn More
Medicom™ AssureMask™ Balance™ Earloop Face Masks
Catalog No. 17000545
Change view
Click to view available options
Color:
Blue
Pink
White
Yellow
ASTM Level:
ASTM Level 2
ASTM Level 3
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color ASTM Level
17000545 White ASTM Level 2
19910740 Blue ASTM Level 2
17000546 Pink ASTM Level 2
17000271 Yellow ASTM Level 2
17000544 Blue ASTM Level 3
17000548 Pink ASTM Level 3
17000547 White ASTM Level 3
17000549 Yellow ASTM Level 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17000545 Supplier Medicom™ Supplier No. 205514
Only null left
Face masks provide healthcare professionals and other frontline workers with the ideal balance between protection and comfort without gapping or irritation.
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • Adjustable nosepiece ensures a comfortable, customized fit
  • Ultrasonically sealed, soft, stretchy earloops are free of natural rubber latex, preventing pulling and irritation
  • Made from high-quality materials that provide reliable protection without compromising breathability
  • Assorted colors available
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 2
Color White
Fastening Type Earloops
Material Spunbond Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Disposable Single-use
Latex Free Yes
Model 205514
Packaging Quantity 50 Masks/Box, 10 Boxes/Cs.
Show More Show Less