Learn More
Medicom™ AssureMask Balance™ Procedure Earloop Face Masks, ASTM Level 1
Catalog No. 296368454
Change view
Click to view available options
Color:
Blue
White
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
296368454 White
296368455 Blue
296368457 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 296368454 Supplier Medicom™ Supplier No. 205414
AssureMask Balance™ Procedure Earloop Face Masks, ASTM Level 1 provide healthcare professionals and other frontline workers with the ideal balance between the protection they need and the breathability and comfort they want. Made from high-quality materials.
  • Shingle pleat design prevents fluid pooling and cross-contamination
  • Made from high-quality materials for reliable protection without compromising breathability
  • Adjustable nosepiece ensures a comfortable, customized fit
    Ultrasonically sealed, soft, stretchy, latex-free earloops prevent pulling and irritation
  • Available in all 3 levels of ASTM protection
Specifications
Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color White
Fastening Type Earloops
Sterility Non-sterile
Style Pleated
Disposable Single-use
Latex Free Yes