missing translation for 'onlineSavingsMsg'
Learn More

Aspirin Impurity D Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014482
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014482 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014482 Supplier Merck Emd Millipore Supplier No. PHR194050MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Aspirin Impurity D is an impurity of the antiplatelet drug aspirin, and one of the major antithrombogenic agent widely used for the prevention of heart attacks and stroke.Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 530-75-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C16H12O6
Synonym 2-[[2-(Acetyloxy)benzoyl]oxy]benzoic acid; Acetylsalicylsalicylic acid; 2-Carboxyphenyl o-acetylsalicylate
Type Certified Reference Material
Quantity 50 mg
Formula Weight 300.26
Grade Certified Reference Material