Learn More
United Scientific™ Aspirator
For use with vacuum aspiration and other lab work.
Supplier:  United Scientific™ ASPRT1
Catalog No. S15758
Only null left
Explore available promotions
Description
Description
  • Strong plastic aspirator for vacuum filtration and other lab applications.
  • Designed to prevent any flooding, back-flow, or turbulence.
  • Includes 5.9 in. (15 cm) hose. The top nozzle has an outer diameter of 1.5 cm.
  • The side nozzle can accommodate hoses from 0.5 cm to 1.0 cm inner diameter.
Specifications
Specifications
Vaccum aspiration
Aspirator
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Warning: Cancer and reproductive harm