missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspartame 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Packaging | Glass Bottle |
| Quantity | 25 g |
| Formula Weight | 294.31 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Grade | Laboratory |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Aspartame |
Safety and Handling
| EINECSNumber | (9)-1616 |
| RTECSNumber | WM3407000 |
| TSCA | Yes |