missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Aspartame, 98%

Catalog No. AAJ6152306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6152306 5 g
AAJ6152314 25 g
AAJ6152322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6152306 Supplier Thermo Scientific Chemicals Supplier No. J6152306

missing translation for 'validMainframeMsg'

A non-nutritive sweetener and dipeptide ester about 160 times sweeter than sucrose | CAS: 22839-47-0 | C14H18N2O5 | 294.31 g/mol

Aspartame is a synthetic and low-calorie sweetener and is used as an active ingredient in consumer foods, beverages, soft drinks, breath mints, sugar free chewing gum, cocoa mixes, frozen desserts, chewable vitamin supplements, milk drinks, cerals, pharmaceutical drugs and supplements, tabletop sweeteners and diet sodas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aspartame is a synthetic and low-calorie sweetener and is used as an active ingredient in consumer foods, beverages, soft drinks, breath mints, sugar free chewing gum, cocoa mixes, frozen desserts, chewable vitamin supplements, milk drinks, cerals, pharmaceutical drugs and supplements, tabletop sweeteners and diet sodas.

Solubility
Soluble in formic acid, dimethyl sulfoxide. Sparingly soluble in water and ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 22839-47-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.31
MDL Number MFCD00002724
InChI Key IAOZJIPTCAWIRG-QWRGUYRKSA-N
Synonym aspartame, nutrasweet, asp-phe-ome, asp-phe methyl ester, aspartam, aspartamum, aspartamo, l-aspartyl-l-phenylalanine methyl ester, aspartylphenylalanine methyl ester, dipeptide sweetener
PubChem CID 134601
ChEBI CHEBI:2877
IUPAC Name (3S)-3-amino-4-[[(2S)-1-methoxy-1-oxo-3-phenylpropan-2-yl]amino]-4-oxobutanoic acid
SMILES COC(=O)[C@H](CC1=CC=CC=C1)NC(=O)[C@@H](N)CC(O)=O

Specifications

Melting Point 243°C to 245°C
Color White
Linear Formula HOOCCH2CH(NH2)CONHCH(CH2C6H5)COOCH3
Beilstein 2223850
Merck Index 14,839
Solubility Information Soluble in formic acid,dimethyl sulfoxide. Sparingly soluble in water and ethanol.
Quantity 5 g
Formula Weight 294.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Aspartame
Show More Show Less
EINECSNumber 245-261-3
RTECSNumber WM3407000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only