Alkali Scientific ASI Supreme™ Plain Surface Non-Charged Microscope Slides, Frosted White Writing Area, 20 Inner Packs of 72, 1440/cs
Catalog No. 502336202
ASI™ Scientific Supreme™ Plain Surface Non-Charged Microscope Slides are 75 X 25mm and 1.0mm thick. Their key feature is the frosted end on one side, which makes it easier to label them with pencils or pens than on other microscope glasses, as it is sand-blasted glass and not etched.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ASI™ Scientific Supreme™ Plain Surface Non-Charged Microscope Slides are 75 X 25mm and 1.0mm thick. Their key feature is the frosted end on one side, which makes it easier to label them with pencils or pens than on other microscope glasses, as it is sand-blasted glass and not etched.