missing translation for 'onlineSavingsMsg'
Learn More

Ashton Pumpmatic PUMPMATIC™ Liquid-Dispensing System DFS Item

Rapid, drop-free fluid sampling, transferring, and dispensing

Supplier:  Ashton Pumpmatic ASHTON105

 View more versions of this product

Catalog No. 22246659


Only null left
Explore available promotions

Description

Description

  • Speeds and simplifies fluid sampling — from 1mL to 10mL per sample
  • Serological pipette/dispenser combination offers accuracy of a reusable at the cost of a disposable
  • Controlled rate of sampling and dispensing delivers accurate volumes without drips or spills
  • Hazardous liquids do not need to be closer than arm's length
  • Transparent, totally closed system
  • Features accurate, clearly marked, easily-read graduations
  • Smooth-action syringe and plunger with Luer-Lok™ permit rapid, one-handed sampling and dispensing
  • Lightweight, unbreakable, clear polystyrene

Specifications

Specifications

Polystyrene
Individually Wrapped
5 mL
Single-use
Sterile
0.16 oz.
Sampling/Dispensing Syringe
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions