missing translation for 'onlineSavingsMsg'
Learn More

Ashton Pumpmatic PUMPMATIC™ Liquid-Dispensing System

Catalog No. 22246658
Encompass
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
10 mL
Sterility:
Non-sterile
Sterile
Packaging Type:
Bulk
Individually Wrapped
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Packaging Type
22246658 1 mL Sterile Individually Wrapped
22246659 5 mL Sterile Individually Wrapped
22246661 1 mL Non-sterile Bulk
22246662 5 mL Non-sterile Bulk
22246663 10 mL Non-sterile Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22246658 Supplier Ashton Pumpmatic Supplier No. ASHTON101
Only null left

Rapid, drop-free fluid sampling, transferring, and dispensing

  • Speeds and simplifies fluid sampling — from 1mL to 10mL per sample
  • Serological pipette/dispenser combination offers accuracy of a reusable at the cost of a disposable
  • Controlled rate of sampling and dispensing delivers accurate volumes without drips or spills
  • Hazardous liquids do not need to be closer than arm's length
  • Transparent, totally closed system
  • Features accurate, clearly marked, easily-read graduations
  • Smooth-action syringe and plunger with Luer-Lok™ permit rapid, one-handed sampling and dispensing
  • Lightweight, unbreakable, clear polystyrene

Specifications

Material Polystyrene
Sterility Sterile
Packaging Type Individually Wrapped
Volume (English) 0.03 oz.
Volume (Metric) 1 mL
Type Sampling/Dispensing Syringe
Disposable Single-use