missing translation for 'onlineSavingsMsg'
Learn More

Ascorbyl Palmitate, NF, 95-100.5%, Spectrum™ Chemical

Catalog No. 18600788
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
5 kg
25 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600788 25 g Poly Bottle
18600789 125 g Poly Bottle
18600790 500 g Poly Bottle
18600791 5 kg Poly Bottle
18600792 25 kg Poly Bottle
18600793 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18600788 Supplier Spectrum Chemical Mfg Cor Supplier No. A137225GM
Get Up to $500 in Chemicals with Purchase
View all promotions

A1372, 137-66-6, C22H38O7

Spectrum™ Chemical Ascorbyl Palmitate, NF is often used to promote flow of tableting powders and to prevent their sticking in tablet presses. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 137-66-6
Molecular Formula C22H38O7
Molecular Weight (g/mol) 414.54
MDL Number MFCD00005377
InChI Key NULLRXHRYOXSEW-UHFFFAOYNA-N
IUPAC Name 2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 2%
Packaging Poly Bottle
Quantity 25 g
Specific Rotation +21° to +24°
Percent Purity 95 to 100.5%
Grade NF
Physical Form Powder or Waxy Solid
Chemical Name or Material Ascorbyl Palmitate