missing translation for 'onlineSavingsMsg'
Learn More

Ascorbyl Palmitate, FCC, 95%, Spectrum™ Chemical

Catalog No. 18600784
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
5 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600784 25 g Poly Bottle
18600785 125 g Poly Bottle
18600786 500 g Poly Bottle
18600787 5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18600784 Supplier Spectrum Chemical Mfg Cor Supplier No. A137425GM
Get Up to $500 in Chemicals with Purchase
View all promotions

A1374, 137-66-6, C22H38O7

Spectrum™ Chemical Ascorbyl Palmitate, FCC is used as an antioxidant food additive (E number E304). The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 137-66-6
Molecular Formula C22H38O7
Molecular Weight (g/mol) 414.54
MDL Number MFCD00005377
InChI Key NULLRXHRYOXSEW-UHFFFAOYNA-N
IUPAC Name 2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 2%
Melting Range 107-117°C
Packaging Poly Bottle
Quantity 25 g
Specific Rotation +21° to +24°
Percent Purity 95%
Grade FCC
Physical Form Powder or Waxy Solid
Chemical Name or Material Ascorbyl Palmitate
Show More Show Less