missing translation for 'onlineSavingsMsg'
Learn More

Ascorbyl Glucoside Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012914
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012914 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012914 Supplier Merck Emd Millipore Supplier No. PHR14331G
Only null left

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Ascorbyl Glucoside is used to protect skin from exposure to UV rays.

Pharmaceutical (small molecule)

Specifications

CAS 129499-78-1
Physical Form Solid
Linear Formula C12H18O11
MDL Number MFCD23701380
Synonym Ascorbyl Glucoside
Type Certified Reference Material
Quantity 1 g
Formula Weight 338.26
Grade Certified Reference Material