missing translation for 'onlineSavingsMsg'
Learn More

Ascomycin, 95%

Catalog No. p-7122685
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66751MB 25 mg
AAJ66751MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66751MB Supplier Thermo Scientific Chemicals Supplier No. J66751MB

Internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus. | CAS: 104987-12-4 | C43H69NO12 | 792.02 g/mol

Ascomycin is a potent immunosuppressive agent that could be used as a potential therapeutic agent for autoimmune diseases. Ascomycin is an inhibitor of PP2. It is an internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus. This Certified Spiking Solution™ is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the narrow therapeutic range of these immunosuppressants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ascomycin is a potent immunosuppressive agent that could be used as a potential therapeutic agent for autoimmune diseases. Ascomycin is an inhibitor of PP2. It is an internal standard for LC-MS/MS testing of the immunosuppressants tacrolimus, sirolimus, and everolimus. This Certified Spiking Solution™ is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the narrow therapeutic range of these immunosuppressants.

Solubility
Soluble in DMSO at 65mg/ml; soluble in ethanol at 50mg/m; with warming. Very poorly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 104987-12-4
Molecular Formula C43H69NO12
Molecular Weight (g/mol) 792.02
MDL Number MFCD06198665
InChI Key ZDQSOHOQTUFQEM-NURRSENYSA-N
Synonym ascomycin, immunomycin, unii-auf4u5nsjk, ascomycin fk520, auf4u5nsjk, chembl8597, 15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 8-ethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetram, ascomycin from streptomyces hygroscopicus var. ascomyceticus, 15,19-epoxy-3h-pyrido 2,1-c 1,4 oxaazacyclotricosine-1,7,20,21 4h,23h-tetrone, 5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-5,19-dihydroxy-8-ethyl-3-2-4-hydroxy-3-methoxycyclohexyl-1-methylethenyl-14,16-dimethoxy-4,10,12,18-tetramethyl-, 3s-3r* e 1s*,3s*,4s* ,4s*,19s*,26ar*, ascomycin fk 520, fr 900520, imm
PubChem CID 5282071
ChEBI CHEBI:29582
SMILES CCC1C=C(CC(CC(C2C(CC(C(O2)(C(=O)C(=O)N3CCCCC3C(=O)OC(C(C(CC1=O)O)C)C(=CC4CCC(C(C4)OC)O)C)O)C)OC)OC)C)C

Specifications

Melting Point 156°C to 160°C
Color White
Boiling Point 868°C
Flash Point >110°C (230°F)
Quantity 25 mg
Solubility Information Soluble in DMSO at 65mg/ml; soluble in ethanol at 50mg/m; with warming; Very poorly soluble in water
Formula Weight 790
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Ascomycin
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber KD4185000
TSCA No
storageNote1 −30° to −10°C
Recommended Storage Store at -20°C

RUO – Research Use Only