missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ASB-14 detergent, ≥97% by TLC, Calbiochem™,

Catalog No. 18275025GM
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18275025GM 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18275025GM Supplier MilliporeSigma™ Supplier No. 18275025GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Zwitterionic amidosulfobetaine detergent. Useful for solubilizing proteins visualized by 2D-electrophoresis.

  • Zwitterionic amidosulfobetaine detergent that exhibits better protein extraction properties than CHAPS
  • Useful for solubilizing proteins visualized by 2D-electrophoresis
  • Increased solubilization produced by this detergent enables the identification of previously undetected membrane proteins

Specifications

Physical Form White to off-white solid
Quantity 25 g
Packaging Glass Bottle
Molecular Formula C22H46N2O4S
Percent Purity ≥97% (TLC)