missing translation for 'onlineSavingsMsg'
Learn More

ASB-14, Calbiochem, ≥ 97% by TLC, MilliporeSigma™

Catalog No. 1827505GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1827505GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1827505GM Supplier MilliporeSigma Supplier No. 1827505GM

A zwitterionic amidosulfobetaine detergent useful for solubilizing proteins visualized by 2D-electrophoresis.

Specifications

Color White to Off-white
Physical Form Solid
Quantity 5 g
Packaging Plastic Ampoule
Molecular Formula C22H46N2O4S
Percent Purity ≥97% (TLC)