missing translation for 'onlineSavingsMsg'
Learn More

3B Scientific™ Artificial Blood

Catalog No. S00284
Change view
Click to view available options
Size:
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S00284 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00284 Supplier 3B Scientific™ Supplier No. 1005691

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Although not the same viscosity as real blood, Life/form™ simulation blood provides a sense of reality to any activity.

  • Although not the same viscosity as real blood, Life/form™ simulation blood provides a sense of reality to any activity.
  • For use with Life/form™ Products:
    • Advanced Venipuncture and Injection Arm (W44216, W44217)
    • Venipuncture and Injection Demonstration Arm
    • Deluxe IV Training Arm
    • Venipuncture Training Arm
    • Pediatric Arm (W44715)
    • Pediatric Head (W44101)
    • Central Venous Cannulation Simulator (W44017)
    • Heart Catherization Simulator for Total Parenteral Nutrition and Central Venous Catheterization (W44015)
    • First Aid Arm (W44733)

Specifications

Product Type Artificial Blood
Color Red
For Use With (Equipment) Life/Form™ Advanced Venipuncture and Injection Arm, Venipuncture and Injection Demonstration Arm, Deluxe IV Training Arm, Venipuncture Training Arm, Pediatric Arm, Pediatric Head, Central Venous Cannulation Simulator, Heart Catherization Simulator for Total Parenteral Nutrition and Central Venous Catheterization, First Aid Arm
Size 1 gal.
Weight (English) 0.35 lb.