missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Wide Bore Filtered Pipette Tips
GREENER_CHOICE

Catalog No. 212361A
Change view
Click to view available options
Volume (Metric):
200 μL
1000 μL
Fit for Purpose:
Sterile and PCR Ready
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Fit for Purpose
212361A 200 μL Sterile and PCR Ready
212362C 1000 μL Sterile and PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 212361A Supplier Thermo Scientific™ Supplier No. 2069G
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Wide Bore Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART tips are designed to aid in pipetting for a variety of applications.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetteting for a variety of applications:

Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications

Description ART 200G, Filtered, Sterile, Lift-off Lid Rack
Volume (Metric) 200 μL
Filtered Filtered
Sterility Sterile
Model ART 200G
Length (English) 1.96 in.
Length (Metric) 4.97cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Lift-off Lid Rack
Tip Style Genomic/Wide Orifice
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile and PCR Ready
Show More Show Less