missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Gel Loading Pipette Tips
GREENER_CHOICE

Catalog No. 2123620
Change view
Click to view available options
Volume (Metric):
20 μL
100 μL
Fit for Purpose:
Sterile and PCR Ready
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Fit for Purpose
2123620 20 μL Sterile and PCR Ready
212369 100 μL Sterile and PCR Ready
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2123620 Supplier Thermo Scientific™ Supplier No. 2155P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Gel Loading Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard
  • Gel loading for loading acrylamide or agarose gels

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes

Ergonomic pipette tips are designed for Gilson pipetteman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications

Description ART Gel 20P, Filtered, Sterile, Lift-off Lid Rack
Volume (Metric) 20 μL
Filtered Filtered
Sterility Sterile
Model ART Gel 20P
Length (English) 2.79 in.
Length (Metric) 7.09cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Lift-off Lid Rack
Tip Style Gel Loading
For Use With (Equipment) Universal Fit
Product Line ART
Fit for Purpose Sterile and PCR Ready
Show More Show Less