missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ART™ Barrier Specialty Pipette Tips

Catalog No. 21402725
Change view
Click to view available options
Volume (Metric):
1000 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21402725 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21402725 Supplier Thermo Scientific™ Supplier No. 234005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Perform applications accurately and consistently with these specialty pipette tips, which seal when exposed to potential contaminants.

Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.

Features:

  • Sterile tips are certified free from RNase, DNase and pyrogen
  • Available in sterile, nonsterile and in various packaging options
  • Manufactured according to the industry's highest quality standard

Designed to aid in pipetteting for a variety of applications:

Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel

Gel loading for loading acrylamide or agarose gels

Wide orifice genomic for genomic DNA, fragile cell lines and other viscous materials

Solvent Safe for acids, bases and aggressive organic solvents

Compatible with:

SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettetes

Ergonomic pipette tips are designed for Gilson pipetteman™ pipettetes and Thermo Scientific™ Finnpipettete™ Focus pipettetes

Specifications

Volume (Metric) 1000 μL
Sterility Sterile
Model ART 1000 ERGO-G
Length (English) 2.78 in.
Length (Metric) 7.061cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Rack
Tip Style Ergonomic
For Use With (Equipment) Gilson Pipetman, Thermo Scientific Finnpipette Focus
Product Line ART
Fit for Purpose Sterile and PCR Ready
Show More Show Less